Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonaldehyde bis(dimethyl acetal), 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
5kg
Packaging:
Glass bottle
Plastic drum
102-52-3
C7H16O4
164.2
MFCD00008488
XHTYQFMRBQUCPX-UHFFFAOYSA-N
propane, 1,1,3,3-tetramethoxy,tetramethoxypropane,malonaldehyde bis dimethyl acetal,malonaldehyde tetramethyl acetal,malonaldehyde, bis dimethyl acetal,1,1,3,3-tetramethoxy-propane,malonaldehyde bis dime,1,3,3-tetramethoxypropane,ksc174o0b,propane,1,3,3-tetramethoxy
66019
1,1,3,3-tetramethoxypropane
COC(CC(OC)OC)OC
102-52-3
C7H16O4
164.2
MFCD00008488
XHTYQFMRBQUCPX-UHFFFAOYSA-N
propane, 1,1,3,3-tetramethoxy,tetramethoxypropane,malonaldehyde bis dimethyl acetal,malonaldehyde tetramethyl acetal,malonaldehyde, bis dimethyl acetal,1,1,3,3-tetramethoxy-propane,malonaldehyde bis dime,1,3,3-tetramethoxypropane,ksc174o0b,propane,1,3,3-tetramethoxy
66019
1,1,3,3-tetramethoxypropane
COC(CC(OC)OC)OC
Flash Point 54°C
Density 0.9900g/mL
Color Undesignated
Quantity 100g
Packaging Glass bottle
Boiling Point 183.0°C
Refractive Index 1.4060 to 1.4080
Infrared Spectrum Authentic
Specific Gravity 0.99
Assay Percent Range 99+%
Linear Formula (CH3O)2CHCH2CH(OCH3)2
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in hexane,ether,alcohol
Formula Weight 164.2
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Malonaldehyde bis(dimethyl acetal), 99+%
Show More Show Less

GHS P Statement Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Warning

EINECSNumber : 203-037-2

TSCA : TSCA