Learn More
Malondialdehyde Mouse anti-Non-species specific, Biotin, Clone: 11000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259367B
Catalog No. NBP259367B
Add to cart
Description
Description
Malondialdehyde Monoclonal antibody specifically detects Malondialdehyde in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Malondialdehyde
11000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.