Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
141-82-2
C3H4O4
104.06
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid,dicarboxymethane,carboxyacetic acid,methanedicarboxylic acid,kyselina malonova,usaf ek-695,dicarboxylate,dicarboxylic acid,kyselina malonova czech,propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O
141-82-2
C3H4O4
104.06
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid,dicarboxymethane,carboxyacetic acid,methanedicarboxylic acid,kyselina malonova,usaf ek-695,dicarboxylate,dicarboxylic acid,kyselina malonova czech,propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O
Infrared Spectrum Authentic
Packaging Plastic bottle
Flash Point 172°C
Melting Point 130.0°C to 135.0°C
Color White
Quantity 5kg
Assay Percent Range 99%
Linear Formula HO2CCH2CO2H
Beilstein 02,566
Merck Index 15,5774
Solubility Information Solubility in water: 1400g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 104.06
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Malonic acid
Show More Show Less