Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonic Acid (Crystalline/Reagent), Fisher Chemical

Click to view available options
Quantity:
100g
Packaging:
Glass Bottle
141-82-2
C3H4O4
104.061
MFCD00002707
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid,dicarboxymethane,carboxyacetic acid,methanedicarboxylic acid,kyselina malonova,usaf ek-695,dicarboxylate,dicarboxylic acid,kyselina malonova czech,propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O
141-82-2
C3H4O4
104.061
MFCD00002707
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid,dicarboxymethane,carboxyacetic acid,methanedicarboxylic acid,kyselina malonova,usaf ek-695,dicarboxylate,dicarboxylic acid,kyselina malonova czech,propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O
Melting Range 133°C to 137°C
Quantity 100g
Color White
Density 1.63g/cm³
Identification Pass Test
Packaging Glass Bottle
Melting Point 130°C
Linear Formula HOOCCH2COOH
Formula Weight 104.06
Physical Form Solid
Grade Reagent
Chemical Name or Material Malonic Acid
Show More Show Less