Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malononitrile 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
109-77-3
C3H2N2
66.06
MFCD00001883
CUONGYYJJVDODC-UHFFFAOYSA-N
malononitrile,dicyanomethane,cyanoacetonitrile,malonic dinitrile,methylene cyanide,dicyanmethane,malonodinitrile,malonic acid dinitrile,methylenedinitrile,methane, dicyano
8010
CHEBI:33186
propanedinitrile
C(C#N)C#N
109-77-3
C3H2N2
66.06
MFCD00001883
CUONGYYJJVDODC-UHFFFAOYSA-N
malononitrile,dicyanomethane,cyanoacetonitrile,malonic dinitrile,methylene cyanide,dicyanmethane,malonodinitrile,malonic acid dinitrile,methylenedinitrile,methane, dicyano
8010
CHEBI:33186
propanedinitrile
C(C#N)C#N
Quantity 2.5kg
Packaging Glass bottle
Flash Point 112°C
Specific Gravity 1.04
Infrared Spectrum Authentic
Density 1.0400g/mL
Melting Point 30.0°C to 34.0°C
Color Undesignated
Boiling Point 220.0°C
Assay Percent Range 99%
Linear Formula NCCH2CN
Beilstein 02,589
Fieser 04,317
Merck Index 15,5775
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Formula Weight 66.06
Physical Form Crystalline Low Melting Mass
Percent Purity 99%
Chemical Name or Material Malononitrile, 99%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-703-2

RTECSNumber : OO3150000

TSCA : TSCA