Learn More
MALT1 Mouse anti-Human, Alexa Fluor 647, Clone: 50, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143120AF647
Catalog No. NBP143120AH
Add to cart
Description
Description
MALT1 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
MALT1
50
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MALT1
The immunogen for this antibody was MALT1.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase
Mouse
IgG1
0.1 ml
Apoptosis, Cancer, Tumor Suppressors
Primary
10892