Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALT1 Mouse anti-Human, FITC, Clone: 50, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143120F

Catalog No. NBP143120F


Add to cart

Description

Description

MALT1 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

MALT1
50
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MALT1
The immunogen for this antibody was MALT1.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase
Mouse
IgG1
0.1 ml
Apoptosis, Cancer, Tumor Suppressors
Primary
10892
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit