Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALT1 Rabbit anti-Human, Clone: MT1/3159R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MALT1
Clone MT1/3159R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122475
View Documents
Novus Biologicals
NBP30750110UG
100 μg
Each for $406.52
Add to cart
 
NB122476
View Documents
Novus Biologicals
NBP30750120UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MALT1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase
Human MALT1 recombinant fragment (aa701-808)
Monoclonal
Protein A or G purified
MT1/3159R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Apoptosis, Cancer, Tumor Suppressors
10 mM PBS with 0.05% BSA
10892
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit