Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltitol, 97%, Alfa Aesar™

Click to view available options
Quantity:
50g
250g
1000g
585-88-6
C12H24O11
344.313
MFCD00006600
VQHSOMBJVWLPSR-WUJBLJFYSA-N
maltitol,d-maltitol,maltisorb,malbit,amalti syrup,malti mr,amalty mr 100,amalty,4-o-alpha-d-glucopyranosyl-d-glucitol,unii-d65dg142wk
493591
CHEBI:68428
(2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O
585-88-6
C12H24O11
344.313
MFCD00006600
VQHSOMBJVWLPSR-WUJBLJFYSA-N
maltitol,d-maltitol,maltisorb,malbit,amalti syrup,malti mr,amalty mr 100,amalty,4-o-alpha-d-glucopyranosyl-d-glucitol,unii-d65dg142wk
493591
CHEBI:68428
(2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O
Applications
Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Melting Point 149°C to 152°C
Quantity 1000g
Optical Rotation +107° (c=10 in Water)
Assay Percent Range 0.97
Beilstein 89983
Formula Weight 344.32
Percent Purity 97%
Chemical Name or Material Maltitol

RTECSNumber : LZ4394000

TSCA : TSCA