Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Mouse anti-E. coli, Clone: R29.6, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Maltose Binding Protein
Clone R29.6
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122643
View Documents
Novus Biologicals
NBP30758510UG
100 μg
Each for $352.89
Add to cart
 
NB122644
View Documents
Novus Biologicals
NBP30758520UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in E. coli samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Maltose Binding Protein
0.2 mg/ml
Western Blot, Immunofluorescence
Purified
RUO
10 mM PBS with 0.05% BSA
MOS maltose binding protein fusion protein.
Monoclonal
Protein A or G purified
R29.6
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
E. coli
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit