Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Mouse anti-Non-species specific, DyLight™ 350, Clone: R3.2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP276652UV

Catalog No. NBP276652UV


Add to cart

Description

Description

Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Maltose Binding Protein
R3.2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
Mouse
IgG1
0.1 mL
Primary
This antibody is useful for detection and isolation of recombinant MBP fusion proteins.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
MOS maltose binding protein fusion protein
Protein A purified
Store at 4°C in the dark.
Monoclonal
Nonspecies specific
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit