Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Monohydrate, ≥98%, Affymetrix/USB™

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
6363-53-7
C12H22O11·H2O
MFCD00149343
d-+-maltose monohydrate,unii-dm477ee40d,2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate,beta-maltose monohydrate,d-+-maltosemonohydrate,69-79-4 anhydrous,d +-maltose monohydrate,d-+-maltose monohydrate, puriss.,d-+-maltose monohydrate, analytical standard,d-+-maltose monohydrate, bioxtra
6363-53-7
C12H22O11·H2O
MFCD00149343
d-+-maltose monohydrate,unii-dm477ee40d,2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate,beta-maltose monohydrate,d-+-maltosemonohydrate,69-79-4 anhydrous,d +-maltose monohydrate,d-+-maltose monohydrate, puriss.,d-+-maltose monohydrate, analytical standard,d-+-maltose monohydrate, bioxtra
Quantity 500g
Packaging Plastic bottle
Melting Point ~120°C (decomposition)
Formula Weight 360.31
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material Maltose Monohydrate