Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MaltoseBindingProtein Rabbit anti-E. coli, Clone: SR41-04, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267345
Description
MaltoseBindingProtein Monoclonal antibody specifically detects MaltoseBindingProtein in E. coli samples. It is validated for Western Blot.Specifications
MaltoseBindingProtein | |
SR41-04 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
E. coli, Non-species specific |
Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:5000 | |
Purified | |
malE | |
Full length protein | |
Protein A purified | |
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
Primary | |
948538 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title