Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML2 Mouse anti-Human, Allophycocyanin/Cy7, Clone: MAML2/1302, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254588APCCY7

Catalog No. N25458APCY7


Add to cart

Description

Description

MAML2 Monoclonal antibody specifically detects MAML2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

MAML2
MAML2/1302
Flow Cytometry
Conjugated, Purified
MAML2
Recombinant human full-length MAML2
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
DKFZp686N0150, KIAA1819MGC176701, mam-2, MAM2, MAM-3, MAM3MLL-MAML2, mastermind (Drosophila)-like 2, mastermind-like 2 (Drosophila), mastermind-like protein 2
Mouse
IgG2a
0.1 mL
Primary
84441.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit