Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549460

Catalog No. PA549460

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
Specifications

Specifications

MAML2
Polyclonal
PBS with 0.09% sodium azide
Q8IZL2
Mastermind-like protein 2, Mam-2, MAML2, KIAA1819
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84441
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAML2
Liquid
MAML2
KLH conjugated synthetic peptide between 780-814 amino acids from the Central region of human MAML2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit