Learn More
Aviva Systems Biology Corp MAML3 Antibody - middle region (ARP47640_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAML3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAML3. Alternate names: CAGH3, ERDA3, GDN, MAM-2, MAM2, TNRC3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59619