Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAMLD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552025
Description
Immunogen sequence: KRPCLEDVTL AMGPGAHPST ACAELQVPPL TINPSPAAMG VAGQSLLLEN NPMNGNIMGS PFVVPQTTEV GLKGPTVPYY EKINSVPAVD QELQELLEEL TKIQDPSPNE LDLE Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 25%.
CXorf6 is a protein predicted based on an ORF found in chromosome 14.
Specifications
MAMLD1 | |
Polyclonal | |
Unconjugated | |
MAMLD1 | |
CG1; CXorf6; F18; G630014P10Rik; HYSP2; MAMLD1; mastermind like domain containing 1; mastermind-like domain containing 1; mastermind-like domain-containing protein 1; Protein CG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13495 | |
MAMLD1 | |
Recombinant protein corresponding to Human MAMLD1. Recombinant protein control fragment (Product #RP-91982). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction