Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mammaglobin A Mouse anti-Human, Clone: rMGB/4299, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Mammaglobin A
Clone rMGB/4299
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122072
View Documents
Novus Biologicals
NBP30729920UG
20 μg
Each for $159.82
Add to cart
 
NB122071
View Documents
Novus Biologicals
NBP30729910UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Mammaglobin A
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
4250
IgG2a Kappa
Primary
Store at 4°C.
rMGB/4299
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1
Stable CHO cell line producing antibody against Mammaglobin A.
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit