Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mammaglobin A, Mouse anti-Human, Clone: SPM518, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | Mammaglobin A |
---|---|
Clone | SPM518 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Mammaglobin A | |
0.2 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Q13296 | |
4250 | |
Recombinant full-length human Mammaglobin A protein | |
Primary | |
Store at 4°C. |
SPM518 | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
SCGB2A2 | |
IgG1 κ | |
Protein A or G purified | |
Mammaglobin A is a 93 amino acid glycoprotein with homology to other secretoglobin-uteroglobin family members. It was originally identified as a breast cancer restricted biomarker by differential screening. Mammaglobin related to secretoglobin family that includes human uteroglobin and lipophilin. Mammaglobin A antibody stains cytoplasm of normal breast epithelial cells as well as primary and metastatic breast carcinomas. Mammaglobin expression is absent in prostate, kidney, colon, rectum, small intestine, stomach, pancreas, lung, and thyroid tissues. Mammaglobin may be used as part of an immunohistochemical panel for determination of metastatic breast carcinoma and tumor of unknown primary origin. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title