Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mammaglobin A Rabbit anti-Human, Clone: MGB/4057R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Mammaglobin A
Clone MGB/4057R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122085
View Documents
Novus Biologicals
NBP30730610UG
100 μg
Each for $406.52
Add to cart
 
NB122086
View Documents
Novus Biologicals
NBP30730620UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Mammaglobin A
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
4250
IgG Kappa
Primary
Store at 4°C.
MGB/4057R
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Rabbit
Human
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1
Recombinant full-length human Mammaglobin A protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit