Learn More
BioVision IncSupplier Diversity Partner Mammalian Cell Extraction Kit: Mammalian Cell & Tissue Extraction Kit: Convenient Kit to prepare Cell Lysate and Tissue Homogenate in less than 20 minutes. 500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K269500
The Mammalian Cell Extraction Kit provides optimized cell extraction buffer, protease inhibitor cocktail, and DTT for convenient extraction of mammalian proteins from cultured cells and tissue samples, under nondenaturing conditions. Cell lysate prepared using the kit can be used in a variety of applications, such as enzyme activity assays (e.g., caspase activity assays), Western blot analysis, and others. The entire procedure takes less than 20 minutes. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9662