Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN1A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549549

Catalog No. PA549549


Add to cart

Description

Description

Involved in the maturation of Asn-linked oligosaccharides. Progressively trim alpha-1,2-linked mannose residues from Man(9)GlcNAc(2) to produce Man(5)GlcNAc(2).
Specifications

Specifications

MAN1A2
Polyclonal
PBS with 0.09% sodium azide
O60476
Mannosyl-oligosaccharide 1, 2-alpha-mannosidase IB, 3.2.1.113, Mannosidase alpha class 1A member 2, Processing alpha-1, 2-mannosidase IB, Alpha-1, 2-mannosidase IB, MAN1A2, MAN1B
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10905
Western Blot
Unconjugated
MAN1A2
Liquid
MAN1A2
KLH conjugated synthetic peptide between 342-373 amino acids from the Central region of human MAN1A2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit