Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN1C1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MAN1C1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095216
View Documents
Novus Biologicals
NBP305040100UL
100 μg Each for $320.71
Add to cart
 
NB095217
View Documents
Novus Biologicals
NBP30504020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

MAN1C1 Polyclonal antibody specifically detects MAN1C1 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

MAN1C1
Unconjugated
Rabbit
Stem Cell Markers
PBS with 50% glycerol, pH7.3.
57134
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
1,2-alpha-mannosidase IC, Alpha-1,2-mannosidase IC, EC 3.2.1.113, HMICProcessing alpha-1,2-mannosidase IC, MAN1A3, MAN1C, Mannosidase alpha class 1C member 1, mannosidase, alpha, class 1C, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IC, pp6318
Recombinant fusion protein containing a sequence corresponding to amino acids 44-240 of human MAN1C1 (NP_065112.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit