Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN2A2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548385

Catalog No. PA548385

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, monkey and mouse based on sequence homology.

Catalyzes the first committed step in the biosynthesis of complex N-glycans. It controls conversion of high mannose to complex N-glycans; the final hydrolytic step in the N-glycan maturation pathway.
Specifications

Specifications

MAN2A2
Polyclonal
PBS with 0.09% sodium azide
P49641, Q8BRK9
Alpha-mannosidase 2x, Alpha-mannosidase IIx, Man IIx, Mannosidase alpha class 2A member 2, Mannosyl-oligosaccharide 1, 3-1, 6-alpha-mannosidase, MAN2A2, MANA2X
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140481, 4122
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAN2A2
Liquid
MAN2A2
KLH conjugated synthetic peptide between 767-796 amino acids from the Central region of human MAN2A2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit