Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN2B1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MAN2B1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096326
View Documents
Novus Biologicals
NBP305138100UL
100 μg
Each for $320.71
Add to cart
 
NB096327
View Documents
Novus Biologicals
NBP30513820UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

MAN2B1 Polyclonal antibody specifically detects MAN2B1 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

MAN2B1
Unconjugated
Rabbit
Mouse
alpha B, lysosomal, EC 3.2.1, EC 3.2.1.24, Laman, Lysosomal acid alpha-mannosidase, Mannosidase alpha class 2B member 1, Mannosidase alpha-B, mannosidase, alpha, class 2B, member 1
Recombinant fusion protein containing a sequence corresponding to amino acids 640-850 of human MAN2B1 (NP_000519.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
4125
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit