Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN2B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531781
Description
Recommended positive controls: IMR32, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
MAN2B2 | |
Polyclonal | |
Unconjugated | |
MAN2B2 | |
135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Ab2-450; core-specific lysosomal alpha-1,6-Mannosidase; epididymis-specific alpha-mannosidase; HMG17L3; Kiaa0935; Man2b2; mannosidase 2, alpha B2; mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; mKIAA0935; NHC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y2E5 | |
MAN2B2 | |
Recombinant fragment corresponding to a region within amino acids 598 and 930 of Human MAN2B2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction