(±)-Mandelic Acid, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar A142230E

Quantity 2500g

 View more versions of this product

Catalog No. AAA142230E

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range 99%
    Molecular Formula C8H8O3
    Formula Weight 152.15
    Chemical Name or Material (+/-)-Mandelic acid

    Specifications

    Formula Weight 152.15
    Quantity 2500g
    Assay Percent Range 99%
    TSCA TSCA
    Molecular Formula C8H8O3
    MDL Number MFCD00064250
    CAS 90-64-2
    Beilstein 510011
    Merck Index 14,5717
    Synonym (+/-)-alpha-Hydroxyphenylacetic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    RTECS Number OO6300000
    Melting Point 118° to 121°C
    Chemical Name or Material (+/-)-Mandelic acid
    Assay 99%

    Description

    Applications
    Mandelic acid is used in the pharmaceutical industry as an antibacterial to treat urinary tract infections. It acts as a precursor for making various drugs. It is also used as an oral antibiotic. Mandelic acid peels are widely used for treating damaged skin due to its less penetrating and irritating property. Enantiomeric resolution of (±)-Mandelic acid is performed by using the chiral amine (1R,2S)-(-)-ephedrine followed by acidification to prepare (R)-( -)-Mandelic acid.

    Solubility
    Soluble in water, diethyl ether, ethanol and isopropanol.

    Notes
    Hydrophobic and light sensitive. Incompatible with strong oxidizing agents and reducing agents.

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit