Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mandelic acid hydrazide, 97%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
2443-66-5
C8H10N2O2
166.18
MFCD00038133
FWTGUGVETHVGTL-UHFFFAOYSA-N
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667
73126
2-hydroxy-2-phenylacetohydrazide
C1=CC=C(C=C1)C(C(=O)NN)O
2443-66-5
C8H10N2O2
166.18
MFCD00038133
FWTGUGVETHVGTL-UHFFFAOYSA-N
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667
73126
2-hydroxy-2-phenylacetohydrazide
C1=CC=C(C=C1)C(C(=O)NN)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 128°C to 131°C
Beilstein 2096197
Quantity 25g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Mandelic acid hydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

RTECSNumber : OO7450000

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only