Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mandelonitrile (Tech.), ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
532-28-5
C8H7NO
133.15
MFCD00004487
NNICRUQPODTGRU-UHFFFAOYSA-N
mandelonitrile,benzaldehyde cyanohydrin,amygdalonitrile,phenylglycolonitrile,mandelic acid nitrile,glycolonitrile, phenyl,benzaldehydkyanhydrin,acetonitrile, hydroxyphenyl,nitril kyseliny mandlove,alpha-hydroxybenzeneacetonitrile
10758
CHEBI:16910
2-hydroxy-2-phenylacetonitrile
C1=CC=C(C=C1)C(C#N)O
532-28-5
C8H7NO
133.15
MFCD00004487
NNICRUQPODTGRU-UHFFFAOYSA-N
mandelonitrile,benzaldehyde cyanohydrin,amygdalonitrile,phenylglycolonitrile,mandelic acid nitrile,glycolonitrile, phenyl,benzaldehydkyanhydrin,acetonitrile, hydroxyphenyl,nitril kyseliny mandlove,alpha-hydroxybenzeneacetonitrile
10758
CHEBI:16910
2-hydroxy-2-phenylacetonitrile
C1=CC=C(C=C1)C(C#N)O
Refractive Index 1.5305 to 1.5335 (20°C, 589nm)
Density 1.11g/mL
Quantity 25mL
Color Yellow to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula C6H5CH(OH)CN
Beilstein 10, 193
Merck Index 15, 5783
Formula Weight 133.15
Physical Form Liquid
Percent Purity ≥80% (typically)
Grade Technical
Chemical Name or Material Mandelonitrile, tech.
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-532-7

RTECSNumber : OO8400000

TSCA : TSCA