Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MANEAL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569639
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 79%; Rat: 100%; Zebrafish: 93%.
MANEAL is a protein coding gene. Gene Ontology (GO) annotations related to this gene include hydrolase activity, acting on glycosyl bonds.
Specifications
MANEAL | |
Polyclonal | |
Unconjugated | |
MANEAL | |
glycoprotein endo-alpha-1,2-mannosidase-like protein; Gm50; maneal; mannosidase endo-alpha like; mannosidase, endo-alpha-like; RGD1563973; si:ch211-30b16.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
149175 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VSG8 | |
MANEAL | |
synthetic peptide directed towards the middle region of human MANEAL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction