Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MANEAL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566582
Description
Immunogen sequence: FYYSWYGSPRR EGHYIHWDHV MVPHWDPKIS ASY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
MANEAL is a protein coding gene. Gene Ontology (GO) annotations related to this gene include hydrolase activity, acting on glycosyl bonds.
Specifications
MANEAL | |
Polyclonal | |
Unconjugated | |
MANEAL | |
glycoprotein endo-alpha-1,2-mannosidase-like protein; Gm50; maneal; mannosidase endo-alpha like; mannosidase, endo-alpha-like; RGD1563973; si:ch211-30b16.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
149175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VSG8 | |
MANEAL | |
Recombinant Human MANEAL. Recombinant protein control fragment (Product #RP-107234). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction