Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100mL
150mL
Packaging:
Narrow Mouth Poly Bottle
Poly Bottle

Chemical Identifiers

Mn
Manganese, 1000μg/mL (0.10% w/v)
BAKER INSTRA-ANALYZED™ Reagent

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Molecular Formula Mn
Formula Weight 54.94
Packaging Narrow Mouth Poly Bottle
Quantity 150mL
Specific Gravity 1L = 1.01kg
Chemical Name or Material Manganese, 1000μg/mL (0.10% w/v)
Grade BAKER INSTRA-ANALYZED™ Reagent