Alfa Aesar™ Manganese aluminide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 3638414

Quantity 25g

 View more versions of this product

Catalog No. AA3638414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12253-13-3
    Formula Weight 135.88
    MDL Number MFCD00213943
    Chemical Name or Material Manganese aluminide

    Specifications

    Physical Form Lumps
    Formula Weight 135.88
    TSCA TSCA
    Particle Size 10mm & down
    MDL Number MFCD00213943
    CAS 12253-13-3
    Quantity 25g
    Chemical Name or Material Manganese aluminide
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit