Alfa Aesar™ Manganese antimonide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 3639014

Quantity 25g

 View more versions of this product

Catalog No. AA3639014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12032-97-2
    Linear Formula Mn2Sb
    Formula Weight 231.63
    MDL Number MFCD00151354
    UN Number UN1549
    Chemical Name or Material Manganese antimonide

    Specifications

    Physical Form Lumps
    Formula Weight 231.63
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula Mn2Sb
    CAS 12032-97-2
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Particle Size 10mm & down
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
    TSCA TSCA
    MDL Number MFCD00151354
    UN Number UN1549
    Quantity 25g
    Health Hazard 1 Warning
    Chemical Name or Material Manganese antimonide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit