Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn

Manufacturer:  Alfa Aesar™ 38607A1

Quantity 1kg

 View more versions of this product

Catalog No. AA38607A1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13434-24-7
MFCD00058698
Manganese(II) 2-ethylhexanoate
341.35
UN1268

Specifications

40% w/w in min.eral spirits, 6% Mn
Petroleum-like
1kg
0.897g/mL
341.35
UN1268
40° (104°F)
Brown
13434-24-7
Liquid
MFCD00058698
Manganese(II) 2-ethylhexanoate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H304-H361-H226
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit