Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Manganese(II) acetate, anhydrous, 98+%

Click to view available options
Quantity:
50g
250g
1000g
638-38-0
C4H6MnO4
173.026
MFCD00013039
UOGMEBQRZBEZQT-UHFFFAOYSA-L
manganese ii acetate,manganese acetate,manganous acetate,diacetylmanganese,manganese diacetate,manganese di acetate,manganese 2+ acetate,octan manganaty czech,acetic acid, manganese 2+ salt,unii-0v6e9q2i0y
12525
manganese(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Mn+2]
638-38-0
C4H6MnO4
173.026
MFCD00013039
UOGMEBQRZBEZQT-UHFFFAOYSA-L
manganese ii acetate,manganese acetate,manganous acetate,diacetylmanganese,manganese diacetate,manganese di acetate,manganese 2+ acetate,octan manganaty czech,acetic acid, manganese 2+ salt,unii-0v6e9q2i0y
12525
manganese(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Mn+2]
Applications
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.

Solubility
Soluble in water, methanol and acetic acid.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
Quantity 250g
Linear Formula Mn(OOCCH3)2
Merck Index 14,5724
Formula Weight 173.03
Percent Purity ≥98%
Chemical Name or Material Manganese(II) acetate, anhydrous

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AI5770000

TSCA : TSCA