Learn More
Manganese(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
Description
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.
Solubility
Soluble in water, methanol and acetic acid.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Linear Formula | Mn(OOCCH3)2 |
Quantity | 50 g |
Sensitivity | Hygroscopic |
Merck Index | 14,5724 |
Solubility Information | Soluble in water,methanol and acetic acid. |
Formula Weight | 173.03 |
Percent Purity | ≥98% |
Chemical Name or Material | Manganese(II) acetate, anhydrous |
Safety and Handling
Hazard Category | H315-H319-H335-H336-H373 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c |
EINECSNumber | 211-334-3 |
RTECSNumber | AI5770000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.