Learn More
Manganese(II) acetate tetrahydrate, Mn 22% (typical), Thermo Scientific Chemicals
Description
It is used as a mordant in dyeing, drier for paints and varnishes, an oxidation catalyst, and in the manufacture of bister. It is also used in the manufacture of fertilizer. It is used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
Specifications
Melting Point | 80°C |
Color | Pink |
Density | 1.589 g/mL |
Odor | Pungent |
Linear Formula | Mn(OOCCH3)2·4H2O |
Quantity | 2 kg |
Merck Index | 14,5724 |
Solubility Information | Soluble in water,methanol,ethanol and acetic acid. |
Formula Weight | 245.09 (173.03 Anhydrous) |
Concentration or Composition (by Analyte or Components) | Mn 22% (typical) |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335-H336-H373 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c |
EINECSNumber | 211-334-3 |
RTECSNumber | AI5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.