Alfa Aesar™ Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1080214

Quantity 25g

 View more versions of this product

Catalog No. AA1080214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6156-78-1
    Linear Formula Mn(OOCCH3)2·4H2O
    Formula Weight 245.09 (173.03 Anhydrous)
    MDL Number MFCD00062552
    Merck Index 14,5724
    Chemical Name or Material Manganese(II) acetate tetrahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 245.09 (173.03 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Pungent
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Physical Form Crystalline Aggregates
    MDL Number MFCD00062552
    Melting Point 80°C
    Density 1.589g/mL
    Linear Formula Mn(OOCCH3)2·4H2O
    CAS 6156-78-1
    Merck Index 14,5724
    Color Pale pink
    Health Hazard 1 Warning
    RTECS Number AI5775000
    Chemical Name or Material Manganese(II) acetate tetrahydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit