Alfa Aesar™ Manganese(II) bromide hydrate, 98%

Manufacturer: Alfa Aesar™ 4024536

Quantity 500g

 View more versions of this product

Catalog No. AA4024536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10031-20-6
    Formula Weight 214.76 (Anhydrous)
    MDL Number MFCD00149791
    Merck Index 14,5725
    Chemical Name or Material Manganese(II) bromide hydrate

    Specifications

    Formula Weight 214.76 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Pink
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Sensitivity Hygroscopic
    MDL Number MFCD00149791
    Melting Point 64.3°C (dec.)
    Health Hazard 1 Warning
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline Aggregates
    CAS 10031-20-6
    Merck Index 14,5725
    Quantity 500g
    Solubility Information Soluble in water, alcohol
    Chemical Name or Material Manganese(II) bromide hydrate
    Assay 98%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit