Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) carbonate, Mn 44% min., Alfa Aesar™

For Pigment, Varnish Drier.

Manufacturer:  Alfa Aesar 1432436

Quantity 500g

 View more versions of this product

Catalog No. AA1432436


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

598-62-9
Mn 44% min.
114.95
Manganese(II) carbonate
Powder
CMnO3
MFCD00011116

Specifications

598-62-9
Mn 44% min.
Light brown to violet powder
MFCD00011116
Light brown to violet
Manganese(II) carbonate
Powder
CMnO3
114.95
Odorless
500g
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit