Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Manganese(II) chloride, 1M aq. soln.

Click to view available options
Quantity:
50mL
100mL
7773-01-5
Cl2H8MnO4
197.90
MFCD00011114
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese chloride,manganese ii chloride,manganese 2+ ion dichloride,manganesechloride,acmc-20akkt,manganous chloride,anhydrous,ksc171o6h,manganous chloride, anhydr,manganese ii chloride anhydrous crystalline
10313134
CHEBI:63041
manganese(2+);dichloride
[Cl-].[Cl-].[Mn+2]
7773-01-5
Cl2H8MnO4
197.90
MFCD00011114
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese chloride,manganese ii chloride,manganese 2+ ion dichloride,manganesechloride,acmc-20akkt,manganous chloride,anhydrous,ksc171o6h,manganous chloride, anhydr,manganese ii chloride anhydrous crystalline
10313134
CHEBI:63041
manganese(2+);dichloride
[Cl-].[Cl-].[Mn+2]
Linear Formula MnCl2
Formula Weight 125.84
Physical Form Liquid
Concentration 1M aq. soln.
Color Colorless
Quantity 100mL
Chemical Name or Material Manganese(II) chloride

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA