Alfa Aesar™ Manganese(II) chloride tetrahydrate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4444222

Quantity 100g

 View more versions of this product

Catalog No. AA4444222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13446-34-9
    Molecular Formula MnCl2·4H2O
    Formula Weight 197.90 (125.84 Anhydrous)
    MDL Number MFCD00149792
    Merck Index 14,5728
    Chemical Name or Material Manganese (II) chloride tetrahydrate

    Specifications

    Formula Weight 197.90 (125.84 Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    Merck Index 14,5728
    Quantity 100g
    TSCA TSCA
    Molecular Formula MnCl2·4H2O
    MDL Number MFCD00149792
    CAS 13446-34-9
    Density 2.010g/mL
    Odor Odorless
    Physical Form Crystalline
    Melting Point 58°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material Manganese (II) chloride tetrahydrate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit