Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(Ii) Chloride Tetrahydrate, 99.99% trace metals basis, Honeywell™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
Poly Bottle
13446-34-9
MFCD00149792
13446-34-9
MFCD00149792

99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Linear Formula MnCl2 · 4H2O
UN Number NONH for all modes of transport
Appearance Crystals
Formula Weight 197.91g/mol
Percent Purity 99.99% trace metals basis
Density 1.913g/cu.cm
Packaging Glass Bottle
Color Pink
Melting Point 58°C
Quantity 5g
Chemical Name or Material Manganese(Ii) Chloride Tetrahydrate
Show More Show Less

P273

H302-H412

EINECSNumber : 231-869-6

RTECSNumber : OO9650000