Alfa Aesar™ Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1446414

Quantity 25g

 View more versions of this product

Catalog No. AA1446414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7773-01-5
    Linear Formula MnCl2
    Formula Weight 125.84
    MDL Number MFCD00011114
    UN Number UN3288
    Merck Index 14,5728
    Chemical Name or Material Manganese(II) chloride, Ultra dry

    Specifications

    Formula Weight 125.84
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    CAS 7773-01-5
    Melting Point 650°C
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Storage Note 1
    • Ampouled under argon
    Mesh Size −10 Mesh
    Physical Form Beads
    Sensitivity Hygroscopic
    UN Number UN3288
    Boiling Point 1190°C
    Quantity 25g
    MDL Number MFCD00011114
    Merck Index 14,5728
    Density 2.977g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Color Pink
    TSCA TSCA
    Linear Formula MnCl2
    Health Hazard 1 Danger
    RTECS Number OO9625000
    Chemical Name or Material Manganese(II) chloride, Ultra dry
    Assay 99.99%, (metals basis)

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit