Learn More
Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1190°C |
Melting Point | 650°C |
Color | Pink |
Physical Form | Beads |
Quantity | 5 g |
Linear Formula | MnCl2 |
UN Number | UN3288 |
Merck Index | 14,5728 |
Solubility Information | Soluble in water(723g/L at 25 C) |
IUPAC Name | manganese(2+);dichloride |
Show More |
Safety and Handling
Hazard Category | H302-H318-H373 |
Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
Precautionary Statement | P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 231-869-6 |
RTECSNumber | OO9625000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.