Alfa Aesar™ Manganese(II) hexafluoro-2,4-pentanedionate trihydrate, 97%

Manufacturer: Alfa Aesar™ H5509403

Quantity 1g

 View more versions of this product

Catalog No. AAH5509403

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 123334-26-9
    Formula Weight 523.09 (469.04 Anhydrous)
    MDL Number MFCD00150654
    Synonym Manganese(II) hexafluoroacetylacetonate trihydrate
    Chemical Name or Material Manganese(II) hexafluoro-2,4-pentanedionate trihydrate

    Specifications

    Formula Weight 523.09 (469.04 Anhydrous)
    MDL Number MFCD00150654
    Synonym Manganese(II) hexafluoroacetylacetonate trihydrate
    Color Yellow
    Melting Point 173°C (dec.)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev
    Physical Form Powder
    CAS 123334-26-9
    Quantity 1g
    Chemical Name or Material Manganese(II) hexafluoro-2,4-pentanedionate trihydrate
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit