Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
It is often used in the lighting industry and it is a source of its derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Melting Point | 638°C |
Color | Red to White |
Physical Form | Beads |
Quantity | 10 g |
Merck Index | 14,5732 |
Solubility Information | Soluble in water. |
Formula Weight | 308.75 |
Percent Purity | 99.99% |
Odor | Odorless |
Packaging | Ampule |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
Precautionary Statement | GHS P Statement P264-P270-P301+P312-P330-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 232-201-6 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction