Manganese(II) nitrate, Alfa Aesar™

Manufacturer: Alfa Aesar 3334018

Quantity 50g

 View more versions of this product

Catalog No. AA3334018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10377-66-9
    Physical Form Liquid
    Molecular Formula MnN2O6
    Formula Weight 178.95
    MDL Number MFCD00011112
    Chemical Name or Material Manganese(II) nitrate, 50% w/w aq. Soln

    Specifications

    Formula Weight 178.95
    Quantity 50g
    Color Pink
    DOT Information DOT Class: 5.1; Packing Group: III
    EINECS Number 233-828-8
    Physical Form Liquid
    Molecular Formula MnN2O6
    Appearance Pink liquid
    MDL Number MFCD00011112
    CAS 10377-66-9
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Health Hazard 1 H272-H351-H315-H319-H335: May intensify fire; oxidizer. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Manganese(II) nitrate, 50% w/w aq. Soln

    H272-H351-H315-H319-H335: May intensify fire; oxidizer. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit