Manganese(II) nitrate hydrate, 99.98% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8784836

Quantity 500g

 View more versions of this product

Catalog No. AA8784836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15710-66-4
    Physical Form Lump
    Assay Percent Range 99.98% (metals basis)
    Molecular Formula Mn(NO3)2·xH2O
    Formula Weight 178.95
    MDL Number MFCD00149788
    Chemical Name or Material Manganese(II) nitrate hydrate

    Specifications

    Formula Weight 178.95
    Physical Form Lump
    Quantity 500g
    Color Pink
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 233-828-8
    Odor Odorless
    Assay Percent Range 99.98% (metals basis)
    Molecular Formula Mn(NO3)2·xH2O
    MDL Number MFCD00149788
    CAS 15710-66-4
    Health Hazard 1 H272-H301-H332-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Manganese(II) nitrate hydrate

    H272-H301-H332-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit